Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTFMT Mouse anti-Human, Clone: OTI1E7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503565

Catalog No. 50-167-4426


Add to cart

Description

Description

The protein encoded by this nuclear gene localizes to the mitochondrion, where it catalyzes the formylation of methionyl-tRNA.
Specifications

Specifications

MTFMT
Monoclonal
0.44 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96DP5
COXPD15, FMT1
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
123263
Immunohistochemistry (Paraffin), Western Blot
OTI1E7
Unconjugated
MTFMT
Liquid
MTFMT
Human recombit protein fragment corresponding to amino acids 175-389 of human MTFMT produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit