Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTFMT Mouse anti-Human, Clone: OTI1E7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525687

Catalog No. PIMA525687


Add to cart

Description

Description

The protein encoded by this nuclear gene localizes to the mitochondrion, where it catalyzes the formylation of methionyl-tRNA.
Specifications

Specifications

MTFMT
Monoclonal
0.44 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96DP5
MTFMT, COXPD15, FMT1
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
123263
Immunohistochemistry (Paraffin), Western Blot
OTI1E7
Unconjugated
MTFMT
Liquid
MTFMT
Human recombit protein fragment corresponding to amino acids 175-389 of MTFMT produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit