Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTHFS Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549053

Catalog No. PA549053

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Contributes to tetrahydrofolate metabolism. Helps regulate carbon flow through the folate-dependent one-carbon metabolic network that supplies carbon for the biosynthesis of purines, thymidine and amino acids.
Specifications

Specifications

MTHFS
Polyclonal
PBS with 0.09% sodium azide
P49914, Q9D110
5-formyltetrahydrofolate cyclo-ligase, 10-methenyl-tetrahydrofolate synthetase, MTHFS, Methenyl-THF synthetase, MTHFS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10588, 107885
Western Blot
Unconjugated
MTHFS
Liquid
MTHFS
KLH conjugated synthetic peptide between 133-159 amino acids from the C-terminal region of human MTHFS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit