Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTHFSD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541900
Description
Peptide sequence: EVKVDPDKPL EGVRLLVLQS KKTLLVPTPR LRTGLFNKIT PPPGATKDIL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 90%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 92%; Rat: 93%; Zebrafish: 77%.
MTHFSD (Methenyltetrahydrofolate Synthetase Domain Containing) is a Protein Coding gene. Diseases associated with MTHFSD include Pancreas, Annular and Anus, Imperforate. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and nucleotide binding. [GeneCards].
Specifications
MTHFSD | |
Polyclonal | |
Unconjugated | |
MTHFSD | |
AW049977; BC052066; C920004D05; fc43c12; LOW QUALITY PROTEIN: methenyltetrahydrofolate synthase domain-containing protein; methenyltetrahydrofolate synthase domain-containing protein; methenyltetrahydrofolate synthase domain-containing protein; LOW QUALITY PROTEIN: methenyltetrahydrofolate synthase domain-containing protein; methenyltetrahydrofolate synthetase domain containing; methenyltetrahydrofolate synthetase domain-containing protein; MGC137477 protein; Mthfsd; si:dkey-199i24.1; Unknown (protein for MGC:153374); wu:fc43c12; zgc:153374 | |
Rabbit | |
Protein A | |
RUO | |
64779 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2M296 | |
MTHFSD | |
Synthetic peptide directed towards the N-terminal of human MTHFSD (aa 61-110). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction