Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTMR2 Mouse anti-Human, Clone: OTI2D2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502140

Catalog No. 50-167-3220


Add to cart

Description

Description

This gene is a member of the myotubularin family. The encoded protein possesses phosphatase activity towards phosphatidylinositol-3-phosphate and phosphatidylinositol-3,5-bisphosphate. Mutations in this gene are a cause of Charcot-Marie-Tooth disease type 4B, an autosomal recessive demyelinating neuropathy. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications

Specifications

MTMR2
Monoclonal
0.9 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13614
CMT4B, CMT4B1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8898
Flow Cytometry, Western Blot
OTI2D2
Unconjugated
MTMR2
Liquid
MTMR2
Full length human recombit protein of human MTMR2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit