Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTMR2 Mouse anti-Human, Mouse, Rat, Canine, Monkey, DyLight 755, Clone: OTI1F10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272810IR
Description
MTMR2 Monoclonal specifically detects MTMR2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.Specifications
MTMR2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MTMR2 | |
Full length recombinant protein of human MTMR2 (NP_057240) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot | |
OTI1F10 | |
Western Blot | |
CMT4B, CMT4B1, EC 3.1.3, EC 3.1.3.2, EC 3.1.3.48, KIAA1073EC 3.1.3.-, myotubularin related protein 2, myotubularin-related protein 2 | |
Mouse | |
Affinity Purified | |
Cancer, Protein Phosphatase | |
8898.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title