Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mTOR/FRAP Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Supplier: Bioss BS1992R
Description
Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.Specifications
mTOR/FRAP | |
Polyclonal | |
Unconjugated | |
MTOR | |
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; Frap; FRAP1; FRAP2; Mammalian target of rapamycin; Mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; Rapamycin target protein 1; RAPT1; Serine/threonine-protein kinase mTOR; SKS | |
Rabbit | |
Protein A | |
RUO | |
2475 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P42345 | |
MTOR | |
KLH conjugated synthetic peptide derived from human mTOR. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction