Learn More
MTOR Mouse anti-Human, Clone: OTI10A2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806066
Catalog No. 50-166-7679
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mammalian target of rapamycin (mTOR) is a serine/threonine kinase that plays a key role in cell growth, cell proliferation, and protein synthesis. mTOR mediates phosphoinositide 3-kinase and Akt/PKB signaling, resulting in phosphorylation of 4EBP1, and initiation of mRNA translation. A second pathway involves regulation of ribosomal S6 kinase, which affects ribosome biogenesis and translation elongation. Mutations affecting the gene results in Smith-Kingsmore syndrome.
Specifications
Specifications
MTOR
Monoclonal
Unconjugated
MTOR
Lyophilized
MTOR
Human recombit protein fragment corresponding to amino acids 1766-2144 of human MTOR produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI10A2
PBS with 8% trehalose and no preservative; pH 7.3
P42345
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2475