Learn More
MTPAP Mouse anti-Human, Clone: 3D2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527842
Catalog No. PIMA527842
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Positive Control: U87-MG, SK-N-SH, IMR32 Predicted Reactivity: Mouse (100%), Zebrafish (85%), Xenopus laevis (93%), Pig (100%), Chicken (96%), Rhesus Monkey (100%), Bovine (100%)
mtPAP encodes a protein that is a member of the DNA polymerase type-B-like family. This enzyme synthesizes the 3apos poly(A) tail of mitochondrial transcripts and plays a role in replication-dependent histone mRNA degradation.
Specifications
Specifications
MTPAP
Monoclonal
1.32 mg/ml
PBS with no preservative
Q9NVV4
FLJ10486, mtPAP, RP11305E6.3, PAPD1, RP11-305E6.3
Mouse
IgG2b
100 μl
-20° C, Avoid Freeze/Thaw Cycles
Primary
55149
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
3D2
Unconjugated
MTPAP
Liquid
MTPAP
Human mtPAP
Protein G
RUO
Antibody
Monoclonal
Human