Learn More
Invitrogen™ MTR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114366
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human K562 whole cell. IHC: human placenta tissue, mouse brain tissue, rat brain tissue, rat brain tissue, human pancreatic cancer tissue. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
MTR encodes the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G.
Specifications
MTR | |
Polyclonal | |
Unconjugated | |
MTR | |
5-methyltetrahydrofolate-homocysteine methyltransferase; 5-methyltetrahydrofolate--homocysteine methyltransferase; 5-methyltetrahydrofolate-homocysteine methyltransferase 1; AI894170; cblG; cobalamin-dependent methionine synthase; D830038K18Rik; FLJ33168; FLJ43216; FLJ45386; HMAG; methionine synthase; Methionine synthase-like protein; methioninesynthase; MS; MTR; Vitamin-B12 dependent methionine synthase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
238505, 4548, 81522 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
A6H5Y3, Q99707, Q9Z2Q4 | |
MTR | |
E.coli-derived human MTR recombinant protein (Position: V880-D1095). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.