Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTRF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548863

Catalog No. PA548863

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and mouse based on sequence homology.

Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain non-cognate termination stop codons AGG and AGA.
Specifications

Specifications

MTRF1
Polyclonal
PBS with 0.09% sodium azide
O75570
Peptide chain release factor 1, mitochondrial, MRF-1, MtRF-1, MTRF1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9617
Western Blot
Unconjugated
MTRF1
Liquid
MTRF1
KLH conjugated synthetic peptide between 232-259 amino acids from the Central region of human MTRF1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit