Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTRF1L Mouse anti-Human, PE, Clone: OTI1D11, Novus Biologicals™

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP272814PE

Catalog No. NBP272814PE


Add to cart

Description

Description

MTRF1L Monoclonal specifically detects MTRF1L in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

MTRF1L
Monoclonal
PE
PBS with 0.05% Sodium Azide
MTRF1L
Full length recombinant protein of human MTRF1L (NP_061914) produced in HEK293T cells.
0.1 mL
54516.0
Store at 4°C in the dark.
Flow Cytometry
OTI1D11
Flow Cytometry
mitochondrial translational release factor 1-likemitochondrial, MRF1L, MTRF1A
Mouse
Affinity Purified
Primary
Human
IgG1
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit