Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549560

Catalog No. PA549560

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This protein binds preferentially and cooperatively to ss-DNA. Probably involved in mitochondrial DNA replication. SSBP is a housekeeping gene involved in mitochondrial biogenesis (Tiranti et al.. 1995).[supplied by OMIM].
Specifications

Specifications

SSBP1
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q04837, Q9CYR0, P28042
Single-stranded DNA-binding protein, mitochondrial, Mt-SSB, MtSSB, PWP1-interacting protein 17, SSBP1, SSBP
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
381760, 54304, 6742
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
SSBP1
Liquid
SSBP1
synthetic peptide corresponding to 85-113aa of human SSBP
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit