Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTT, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
500mg
1g
5g
10g
Packaging:
Plastic bottle
298-93-1
C18H16BrN5S
414.325
MFCD00011964
AZKSAVLVSZKNRD-UHFFFAOYSA-M
thiazolyl blue,thiazolyl blue tetrazolium bromide,mmt tetrazolium,methylthiazoletetrazolium,mtt,thiazolyl blue monotetrazolium,thiazole blue,3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide,mtt van,unii-euy85h477i
64965
CHEBI:53233
2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide
CC1=C(SC(=N1)[N+]2=NC(=NN2C3=CC=CC=C3)C4=CC=CC=C4)C.[Br-]
298-93-1
C18H16BrN5S
414.325
MFCD00011964
AZKSAVLVSZKNRD-UHFFFAOYSA-M
thiazolyl blue,thiazolyl blue tetrazolium bromide,mmt tetrazolium,methylthiazoletetrazolium,mtt,thiazolyl blue monotetrazolium,thiazole blue,3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide,mtt van,unii-euy85h477i
64965
CHEBI:53233
2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide
CC1=C(SC(=N1)[N+]2=NC(=NN2C3=CC=CC=C3)C4=CC=CC=C4)C.[Br-]
  • Used in quantitation of DNA synthesis
  • Detects living and growing cells
  • MTT is cleaved by active mitochondria to form dark blue formazan product
Quantity 1g
Packaging Plastic bottle
Melting Point ~195°C (decomposition)
Formula Weight 414.32
Physical Form Powder
Grade Ultrapure
Chemical Name or Material MTT