Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTUS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | MTUS1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP160097
|
Novus Biologicals
NBP160097 |
100 μL |
Each of 1 for $436.00
|
|
Description
MTUS1 Polyclonal specifically detects MTUS1 in Human samples. It is validated for Western Blot.Specifications
MTUS1 | |
Polyclonal | |
Purified | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Angiotensin-II type 2 receptor-interacting protein, AT2 receptor-binding protein, AT2R binding protein, ATBPATIP1, ATIP, DKFZp586D1519, erythroid differentiation-related, FLJ14295, GK1, KIAA1288DKFZp686F20243, microtubule associated tumor suppressor 1, microtubule-associated tumor suppressor 1, Mitochondrial tumor suppressor 1, mitochondrial tumor suppressor gene 1, MP44, MTSG1AT2 receptor-interacting protein, transcription factor MTSG1 | |
MTUS1 | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Breast Cancer | |
Q9ULD2-3 | |
57509 | |
Synthetic peptides corresponding to MTUS1(mitochondrial tumor suppressor 1) The peptide sequence was selected from the N terminal of MTUS1. Peptide sequence QLLACGNTKFEALTVVIQHLLSEREEALKQHKTLSQELVNLRGELVTAST. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title