Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552838
Description
Immunogen sequence: WIDTKNYVEV TRKWYAEAMP FPLNFFLPGR MQRQYMERLQ LLTGEHRPED EEELEKELYR EARECLTLLS QRLGSQKFFF GDAPASLDAF VFSYLALLLQ AKLPSG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
MTX1 belongs to the metaxin family. It is involved in transport of proteins into the mitochondrion and essential for embryonic development.
Specifications
MTX1 | |
Polyclonal | |
Unconjugated | |
MTX1 | |
Gcap6; LOW QUALITY PROTEIN: metaxin-1; Metaxin 1; metaxin-1; Mitochondrial outer membrane import complex protein 1; Mtx; MTX1; MTXN; Unknown (protein for MGC:134321) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4580 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13505 | |
MTX1 | |
Recombinant protein corresponding to Human MTX1. Recombinant protein control fragment (Product #RP-90442). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction