Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MTX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $539.50
Specifications
Antigen | MTX1 |
---|---|
Concentration | 500 μg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119279
|
Invitrogen™
PA5119279 |
200 μL |
Each for $539.50
|
|
|||||
PIPA5121760
|
Invitrogen™
PA5121760 |
20 μL |
Each for $237.00
|
|
|||||
Description
MTX1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
MTX1 belongs to the metaxin family. It is involved in transport of proteins into the mitochondrion and essential for embryonic development.Specifications
MTX1 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Rat | |
MTX1 | |
Gcap6; LOW QUALITY PROTEIN: metaxin-1; Metaxin 1; metaxin-1; Mitochondrial outer membrane import complex protein 1; Mtx; MTX1; MTXN; Unknown (protein for MGC:134321) | |
MTX1 | |
Primary | |
Antigen Affinity Chromatography |
500 μg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
0 | |
295241 | |
Recombinant protein Metaxin 1. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title