Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mu-Calpain Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA126476
Description
PA1-26476 detects Calpain 1 from human samples. PA1-26476 has been successfully used in Western blot procedures.
The calpains, calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1.
Specifications
| Mu-Calpain | |
| Polyclonal | |
| Unconjugated | |
| Q27970, P07384, O35350, P35750, P06815, P97571 | |
| CAPN1 | |
| Antigen affinity chromatography | |
| RUO | |
| 100009089, 12333, 281661, 29153, 397027, 823 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| CAPN1 | |
| CANP, CANP1, CANPL1, muCANP, muCL | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Bovine, Human, Mouse, Pig, Rabbit, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction