Alfa Aesar™ mu-Chloro-mu-methylenebis(cyclopentadienyl)titaniumdimethylaluminum, 0.5M in toluene

Manufacturer: Alfa Aesar™ 44506AA

Quantity 5mL

 View more versions of this product

Catalog No. AA44506AA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67719-69-1
    Molecular Formula C13H18AlClTi
    Formula Weight 284.62
    MDL Number MFCD00151575
    UN Number UN3399
    Merck Index 14,9091
    Synonym Tebbe Reagent
    Chemical Name or Material mu-Chloro-mu-methylenebis (cyclopentadienyl)titaniumdimethylaluminum

    Specifications

    Formula Weight 284.62
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H225-H261-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    Sensitivity Air & Moisture Sensitive
    Physical Form Liquid
    Molecular Formula C13H18AlClTi
    MDL Number MFCD00151575
    Merck Index 14,9091
    Health Hazard 1 Danger
    Density 0.927g/mL
    Quantity 5mL
    UN Number UN3399
    Flash Point 1°C (34°F)
    Color Dark red-brown
    Concentration 0.5M in toluene
    CAS 67719-69-1
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    Synonym Tebbe Reagent
    Chemical Name or Material mu-Chloro-mu-methylenebis (cyclopentadienyl)titaniumdimethylaluminum

    GHS H Statement
    H225-H261-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit