Learn More
BiolegendSupplier Diversity Partner Mouse MDSC Flow Cocktail 1 with Isotype Ctrl  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 147001
Mouse MDSC Flow Cocktail 1 with Isotype Ctrl [M1/70; RB6-8C5; 1A8]; Isotype: Rat IgG2a/Rat IgG2b; Reactivity: Mouse; Apps: FC; Size: 25 tests
Catalog No. 50-711-913