Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mu Opioid Receptor Guinea Pig anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$296.36 - $489.80

Specifications

Antigen Mu Opioid Receptor
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-209-9998 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP10106100UL
100 μL
Each for $489.80
Add to cart
 
50-209-9999 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP1010650UL
50 μL
Each for $296.36
Add to cart
 
Description

Description

Mu Opioid Receptor Polyclonal antibody specifically detects Mu Opioid Receptor in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Specifications

Specifications

Mu Opioid Receptor
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Unconjugated
Guinea Pig
Human, Mouse, Non-human Primate, Rat
P33535, P35372, P42866
18390, 25601, 4988
IgG
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
OPRM1
1 mg/mL
Polyclonal
Liquid, Purified
RUO
PBS with 50% glycerol and no preservative
BOS_9964; hMOP; KIAA0403; LMOR; MOP; MOP receptor; MOP-R; Mor; MOR1; M-OR-1; MOR-1; MOR-1C; MOR-1O; MORA; Mu opiate receptor; Mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; mu opioid receptor splice variant rMOR-1S; mu opioid receptor splice variant rMOR-1Z; mu-opioid receptor; muOR; MUOR1; Mu-type opioid receptor; Opioid mu receptor; Opioid Receptor; Opioid receptor B; opioid receptor mu 1; opioid receptor, mu 1; Oprm; OPRM1; Oprrm1; OTTHUMP00000017463; Ror-b
NHQLENLEAETAPLP
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit