Learn More
Selleck Chemical LLCSupplier Diversity Partner Mubritinib (TAK 165) 25mg 366017-09-6  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S221625MG
Mubritinib (TAK-165) is a potent inhibitor of HER2/ErbB2 with IC50 of 6 nM in BT-474 cell; no activity to EGFR, FGFR, PDGFR, JAK1, Src and Blk in BT-474 cell line. Phase 1.
Catalog No. 50-644-8