Learn More
MUC-4 Mouse anti-Human, Alexa Fluor 405, Clone: 781631, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8195V100UG
Catalog No. FAB8195V100
Add to cart
This item is not returnable. View return policy
Description
Description
MUC4 Monoclonal antibody specifically detects MUC4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
MUC-4
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q99102
MUC4
E. coli-derived recombinant human MUC4 Accession # Q99102
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human MUC4 in ELISA.
Flow Cytometry
781631
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Ascites sialoglycoprotein, ASGP, HSA276359, MUC4, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin
Mouse
IgG2b
RUO
Primary
4585
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)