Learn More
MUC1/CA15-3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 199761AP
Catalog No. 50-173-2633
Add to cart
Description
Description
MUC1 (Mucin 1, Episialin, MAM-6, CA 15-3, PEM and EMA) is a large cell surface mucin glycoprotein expressed by most glandular and ductal epithelial cells and some hematopoietic cell lineages. MUC1 is a transmembrane glycoprotein with a large mucin-like extracellular domain that matures through several intermediate forms generated by proteolysis, and sequential addition and processing of numerous O-linked glycans that are heavily sialylated. MUC1 is highly polymorphic and each allele encodes a product that contains a different number of repeats (between 30 and 90) leading to large differences in molecular weight of the protein. MUC1 is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells, and is expressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands. The MUC1 gene contains seven exons and produces several different alternatively spliced variants. Overexpression, aberrant intracellular localization, and changes in glycosylation of the MUC1 protein have been associated with carcinomas. Multiple alternatively spliced transcript variants that encode different isoforms of the MUC1 gene have been reported, but the full-length nature of only some has been determined. Transgenic MUC-1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands.
Specifications
Specifications
MUC1/CA15-3
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15941
CA15 3, CA153, CA15-3, Carcinoma associated mucin, CD227, EMA, Episialin, H23AG, KL-6, MAM6, MUC 1, MUC1, MUC1 beta, MUC1 CT, MUC1 NT, Mucin 1, Peanut reactive urinary mucin, PEM, PEMT, Polymorphic epithelial mucin, PUM, Tumor associated mucin
Rabbit
IgG
150 μL
-20°C
Primary
4582
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
MUC1
Liquid
MUC1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human