Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Chimeric Recombinant Rabbit Monoclonal Antibody (HMFG2), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA548042

Encompass

Catalog No. PIMA548042


Add to Cart

Description

Description

Specificity: This antibody recognises MUC1, a tumor associated antigen which is expressed in >90% ovarian carcinomas. This antigen is a high molecular weight (M, 80,000-200,000) glycoprotein.

MUC1 Chimeric Recombinant Monoclonal antibody specifically detects MUC1 Chimeric in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MUC1 Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P11049
Rabbit
Affinity chromatography
RUO
951
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy
HMFG2
Unconjugated
CD37
ADMCKD; ADMCKD1; Breast carcinoma-associated antigen DF3; CA 15-3; Cancer antigen 15-3; carcinoma-associated mucin; CD227; DF3 antigen; EMA; Episialin; H23 antigen; H23AG; J19; KL-6; Krebs von den Lungen-6; MAM6; MCD; MCKD; MCKD1; Medullary cystic kidney disease, autosomal dominant; Muc1; MUC-1; MUC-1/SEC; MUC-1/X; MUC1/ZD; MUC1-alpha; MUC1-beta; MUC1-CT; MUC1-NT; mucin 1, cell surface associated; mucin 1, transmembrane; Mucin1; mucin-1; Mucin-1 subunit alpha; Mucin-1 subunit beta; peanut-reactive urinary mucin; PEM; PEMT; Polymorphic epithelial mucin; PUM; tumor associated epithelial mucin; tumor-associated epithelial membrane antigen; Tumor-associated mucin
BALB/c mice were immunized with lymph node cells from a patient with nodular poorly differentiated lymphocytic lymphoma.
200 μg
Primary
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit