Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1 Rabbit anti-Human, Unconjugated, Clone: MUC1/1887R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00580

Catalog No. 89-339-536


Add to cart

Description

Description

MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for IHC-P.
Specifications

Specifications

MUC1
MUC1/1887R
mucin 1, cell surface associated
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Immunohistochemistry (Paraffin)
Unconjugated
MUC1
CD227, EMA, H23AG, MAM6, PEM, PEMT, PUM
Recombinant protein corresponding to human MUC1
100 μg
Primary
4582
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit