Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC15 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548498

Catalog No. PA548498

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May play a role in the cell adhesion to the extracellular matrix.
Specifications

Specifications

MUC15
Polyclonal
Unconjugated
MUC15
Liquid
MUC15
KLH conjugated synthetic peptide between 209-237 amino acids from the C-terminal region of human MUC15.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 0.09% sodium azide
Q8N387
Mucin-15, MUC-15, MUC15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
143662
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit