Learn More
MUC16 Mouse anti-Human, Clone: X75 (3C8/4), Abnova™
Mouse monoclonal antibody raised against native human MUC16.
Manufacturer:  Abnova Corporation MAB12465
Catalog No. 89-934-617
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
MUC16
X75 (3C8/4)
Mouse monoclonal antibody raised against native human MUC16.
In TBS (0.09% sodium azide)
CA125/FLJ14303
Mouse
IgG1
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
MUC16
Native human MUC16.
Ion exchange chromatography purification
RUO
Primary
94025