Learn More
MUC2 Mouse anti-Human, Alexa Fluor™ 350, Clone: CCP58, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233157AF350
Catalog No. NP23317A30
Add to cart
Description
Description
MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for ELISA.
Specifications
Specifications
MUC2
CCP58
ELISA
Intestinal mucin-2, MLP, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, mucin-like protein, SMUC
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Recognizes a single glycoprotein of 520kDa, identified as mucin 2 (MUC2). This MAb shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3. Mucins are high molecular weight glycoproteins, which constitute the major component of the mucus layer that protects the gastric epithelium. MUC2 is specifically expressed in goblet cells of the small intestine & colon; in about 65% of colonic carcinomas and about 40% of gastric carcinomas. MUC2 is rarely expressed outside of the GI tract with the exceptions of mucinous carcinoma of breast and clear cell-type carcinomas of the ovary.
ELISA
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
MUC2
A synthetic peptide of 29 amino acids, KYPTTTPISTTTMVTPTPTPTGTQTPTTT from MUC2 protein, coupled to KLH.
0.1 mL
Primary
4583
Human