Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2 Mouse anti-Human, Clone: MLP/842, Abnova™

Mouse monoclonal antibody raised against full length recombinant human MUC2.

Manufacturer:  Abnova Corporation MAB13368

Catalog No. 89-934-619


Add to cart

Description

Description

This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins produced by many epithelial tissues. The protein encoded by this gene is secreted and forms an insoluble mucous barrier that protects the gut lumen. The protein polymerizes into a gel of which 80% is composed of oligosaccharide side chains by weight. The protein features a central domain containing tandem repeats rich in threonine and proline that varies between 50 and 115 copies in different individuals. Alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq]

Specifications

Specifications

MUC2
MLP/842
Mouse monoclonal antibody raised against full length recombinant human MUC2.
In 10mM PBS.
MLP/SMUC
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This monoclonal antibody shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC3.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
MUC2
Recombinant protein corresponding to full length human MUC2.
Protein A/G purification
RUO
Primary
4583
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit