Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2 Mouse anti-Human, FITC, Clone: 996/1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA528127

Catalog No. PIMA528127


Add to cart

Description

Description

Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.
Specifications

Specifications

MUC2
Monoclonal
1 mg/mL
PBS with 1% BSA and 0.09% sodium azide
Q02817
MUC2
MUC-2 tandem repeat peptide
Protein G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
996/1
FITC
MUC2
Liquid
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Primary
4583
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit