Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2 Mouse anti-Human, KHL, Clone: CCP58, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen MUC2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF19891MI
View Documents
MilliporeSigma
MABF1989100UG
100 μg
Each for $386.00
Add to cart
 
MABF19892MI
View Documents
MilliporeSigma
MABF198925UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

MUC2
KHL
Mouse
MUC-2;Intestinal Mucin-2
KLH-conjugated linear peptide (MI-29) corresponding to 29 amino acids from human Mucin-2 containing one repeat unit of 23 amino acids and part of the next repeat of four amino acids.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ELISA, Immunohistochemistry (Paraffin), Western Blot
Purified
Human
MUC2, SMUC
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit