Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC2 Rabbit anti-Human, Clone: MLP/2970R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen MUC2
Clone MLP/2970R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121950
View Documents
Novus Biologicals
NBP30723820UG
20 μg
Each for $159.82
Add to cart
 
NB121949
View Documents
Novus Biologicals
NBP30723810UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MUC2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC
Recombinant full-length human MUC2 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
MLP/2970R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Cellular Markers, Inflammation, Signal Transduction
10 mM PBS with 0.05% BSA
4583
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit