Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MUC20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA514973
Description
MUC20 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins secreted by many epithelial tissues to form an insoluble mucous barrier. The shorter isoform expressed by this gene is localized to the plasma membrane, whereas the longer isoform might be secreted. The C terminus of this protein associates with the multifunctional docking site of the met proto-oncogene and suppresses activation of some downstream met signaling cascades. The protein features a tandem repeat domain that varies between 2 and 6 copies in most individuals. The allele represented in the human reference assembly contains 12 copies. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
MUC20 | |
Polyclonal | |
Unconjugated | |
Muc20 | |
BC026367; KIAA1359; Muc20; MUC-20; mucin 20; mucin 20, cell surface associated; mucin-20; transmembrane mucin MUC20S; UNQ2782/PRO7170 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
200958 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.8 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8N307 | |
Muc20 | |
KLH conjugated synthetic peptide between 654-684 amino acids from the C-terminal region of human MUC20. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction