Learn More
MUC3 Mouse anti-Human, Clone: M3.1, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human MUC3.
Manufacturer:  Abnova Corporation MAB13369
Catalog No. 89-934-620
Add to cart
Description
Description
The mucin genes encode epithelial glycoproteins, some of which are secreted and some membrane bound. Each of the genes contains at least one large domain of tandemly repeated sequence that encodes the peptide sequence rich in serine and/or threonine residues, which carries most of the O-linked glycosylation (Gendler and Spicer, 1995 [PubMed 7778880]).[supplied by OMIM]
Specifications
Specifications
MUC3
M3.1
Mouse monoclonal antibody raised against synthetic peptide of human MUC3.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
MUC3/mucin
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
This monoclonal antibody shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC2.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
MUC3A
A synthetic peptide (conjugated with KLH) corresponding to a 35 amino acid fragment, SIB35, which contains two tandem repeats of human MUC3.
Protein A/G purification
RUO
Primary
4584/57876
Human