Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC3 Mouse anti-Human, Clone: MUC3/1154, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14365

Catalog No. 89-034-447


Add to cart

Description

Description

Mouse monoclonal antibody raised against synthetic peptide of human MUC3.

  • High specificity
  • High reactivity
Specifications

Specifications

MUC3
MUC3/1154
Mouse monoclonal antibody raised against synthetic peptide of human MUC3.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
MUC3A
A synthetic peptide (conjugated with KLH) corresponding to a 35 amino acid fragment, SIB35, which contains two tandem repeats of human MUC3.
Protein A/G purification
RUO
Primary
4584|57876
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q02505;Q9H195
MUC3|mucin
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
This monoclonal antibody recognizes a protein of HMW, identified as mucin 3 glycoprotein (MUC3). This monoclonal antibody shows no cross-reaction with human milk fat globule membranes, MUC1, or MUC2.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)