Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC3B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548273

Catalog No. PA548273

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Major glycoprotein component of a variety of mucus gels. Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces.
Specifications

Specifications

MUC3B
Polyclonal
PBS with 0.09% sodium azide
Q9H195
Mucin-3B, MUC-3B, Intestinal mucin-3B, MUC3B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57876
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MUC3B
Liquid
MUC3B
KLH conjugated synthetic peptide between 866-895 amino acids from the C-terminal region of human MUC3B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit