Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC4 Antibody (MUC4/3084), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279866F
Description
MUC4 Monoclonal specifically detects MUC4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MUC4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MUC4 | |
Recombinant fragment (around aa 1730-1864) of human MUC4 protein (exact sequence is proprietary) (Uniprot: Q99102) | |
0.1 mL | |
Cancer | |
Human | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MUC4/3084 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction