Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC4, Mouse anti-Human, Alexa Fluor 700, Clone: MUC4/3084, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279866AF700
Description
MUC4 Monoclonal antibody specifically detects MUC4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
MUC4 | |
MUC4/3084 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
MUC4 | |
Recombinant fragment (around aa 1730-1864) of human MUC4 protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Cancer | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title