Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC5AC Monoclonal Antibody (C7), Invitrogen™
Mouse Monoclonal Antibody
$166.00 - $405.00
Specifications
Antigen | MUC5AC |
---|---|
Clone | C7 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543651
![]() |
Thermo Scientific
MA543651 |
100 μL |
Each for $405.00
|
|
PIMA543652
![]() |
Thermo Scientific
MA543652 |
20 μL |
Each for $166.00
|
|
Description
MUC5AC Monoclonal antibody specifically detects MUC5AC in Human samples. It is validated for Western BlotSpecifications
MUC5AC | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
MUC5AC | |
Primary | |
-20°C |
C7 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
MUC5AC | |
P98088 | |
IgG2b κ | |
Protein A/G | |
Antibody |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title