Learn More
MUC5AC Mouse anti-Human, Clone: SPM488, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human MUC5AC.
Manufacturer:  Abnova Corporation MAB13118
Catalog No. 89-934-630
Add to cart
Description
Description
Specifications
Specifications
MUC5AC
SPM488
Mouse monoclonal antibody raised against synthetic peptide of human MUC5AC.
In 10mM PBS.
MUC5
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
MUC5AC
A synthetic peptide corresponding to tandem repeat of human MUC5AC.
Protein A/G purification
RUO
Primary
4586