Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC5AC Antibody (MUC5AC/917), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254448PEATT594
Description
MUC5AC Monoclonal specifically detects MUC5AC in Human, Rat (Negative) samples. It is validated for Flow Cytometry.Specifications
MUC5AC | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
MUC5AC | |
Recombinant fragment covering tandem repeats of human MUC5AC protein (exact sequence is proprietary) (Uniprot: P98088) | |
0.1 mL | |
Primary | |
Mucin 5AC glycoprotein (MUC5AC) is a 641kDa glycoprotein belonging to the superfamily of mucins. Mucins are high molecular weight glycoproteins produced by epithelial cells and can be divided into two families; secretory mucins and membrane bound mucins. MUC5AC is a mucus-forming secreted mucin that is found in normal gastric and tracheo-bronchial mucosa, but absent from normal colon. MUC5AC expression is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastatic to the ovary. MUC5AC antibodies may also be useful for identification of intestinal metaplasia as well as in the identification of pancreatic carcinoma and pre-cancerous changes vs. normal pancreas. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
MUC5AC/917 | |
Flow Cytometry | |
gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin | |
Mouse | |
Protein A or G purified | |
Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction | |
4586.0 | |
Human, Rat (Negative) | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction