Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC5B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5121440

This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA
MUC5B Polyclonal antibody specifically detects MUC5B in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
MUC5B | |
Polyclonal | |
Unconjugated | |
MUC5B | |
Cervical mucin; cervical mucin MUC5B; high molecular weight salivary mucin MG1; MG1; MUC5; MUC5B; MUC-5B; MUC9; mucin 5, subtype B, tracheobronchial; mucin 5B, oligomeric mucus/gel-forming; Mucin-5 subtype B, tracheobronchial; Mucin-5B; sublingual gland mucin | |
Rabbit | |
Protein G | |
RUO | |
727897 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
3.62 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9HC84 | |
MUC5B | |
Recombinant Human Mucin-5B protein (3929-4106aa) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title