Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC6 Mouse anti-Human, Clone: IHC626, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP2759857ML

 View more versions of this product

Catalog No. NBP2759857M

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MUC6 Monoclonal antibody specifically detects MUC6 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications

Specifications

MUC6
IHC626
Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200
gastric mucin, Gastric mucin-6, MUC-6, mucin 6, gastric, mucin 6, oligomeric mucus/gel-forming, mucin-6
Mouse
IgG2a
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Tris Buffer, pH 7.3 - 7.7, with 1% BSA with 0.1% sodium azide
MUC6
Recombinant Human MUC6
7 mL
Primary
4588
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit