Learn More
MUC6 Mouse anti-Human, Clone: SPM598, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human MUC6.
Manufacturer:  Abnova Corporation MAB13119
Catalog No. 89-934-633
Add to cart
Description
Description
Specifications
Specifications
MUC6
SPM598
Mouse monoclonal antibody raised against synthetic peptide of human MUC6.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
MUC6
A synthetic peptide corresponding to tandem repeat of human MUC6.
Protein A/G purification
RUO
Primary
4588
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human