Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC6 Rabbit anti-Human, Clone: MUC6/1553R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen MUC6
Clone MUC6/1553R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 2-4 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122220
View Documents
Novus Biologicals
NBP30737320UG
20 μg
Each for $159.82
Add to cart
 
NB122219
View Documents
Novus Biologicals
NBP30737310UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

MUC6 Monoclonal antibody specifically detects MUC6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MUC6
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
4588
IgG
Primary
Store at 4°C.
MUC6/1553R
Immunohistochemistry-Paraffin 2-4 ug/ml
Unconjugated
Rabbit
Human
gastric mucin, Gastric mucin-6, MUC-6, mucin 6, gastric, mucin 6, oligomeric mucus/gel-forming, mucin-6
Recombinant full-length human MUC6 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit