Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC7 Mouse anti-Human, Janelia Fluor 549, Clone: 4D2-1D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP250391JF549

Catalog No. NBP50391549


Add to cart

Description

Description

MUC7 Monoclonal antibody specifically detects MUC7 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

MUC7
4D2-1D7
Immunohistochemistry
Purified
MUC7
KLH conjugated peptide EGRERDHELRHRRHHHQ located in the N-terminal part of the MUC7 protein within the histatin-like domain.was used to immunise Balb/c female mice.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Myeloma: HPRT-deficient SP2/0. No cross reactivity with other human mucins.
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
apo-MG2, DKFZp686J03256, FLJ27047, MG2MGC34772, MUC-7, mucin 7, secreted, mucin-7, salivary, Salivary mucin-7
Mouse
IgG1 Kappa
0.1mL
Primary
4589
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit