Learn More
Mucin 5AC Mouse anti-Human, Mouse, Rat, Porcine, Chicken, Feline, Primate, Rabbit, Bovine (Negative), Allophycocyanin, Clone: 45M1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP232732APC
Catalog No. NB232732APC
Add to cart
Description
Description
MUC5AC Monoclonal specifically detects MUC5AC in Human, Mouse, Rat, Porcine, Chicken, Feline, Mammal, Monkey, Rabbit, Bovine (Negative) samples. It is validated for Flow Cytometry, Dot Blot.
Specifications
Specifications
Mucin 5AC
45M1
Flow Cytometry
Purified
MUC5AC
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Protein A or G purified
Cellular Markers, Extracellular Matrix, Infections (Virus Bacteria and Parasites), Signal Transduction
Primary
4586
Human, Mouse, Rat, Porcine, Chicken, Feline, Primate, Rabbit, Bovine (Negative)
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
gastric mucin, leB, lewis B blood group antigen, major airway glycoprotein, MUC5, mucin 5, subtypes A and C, tracheobronchial/gastric, mucin 5AC, oligomeric mucus/gel-forming, mucin 5AC, oligomeric mucus/gel-forming pseudogene, mucin-5 subtype AC, tracheobronchial, mucin-5AC, TBM, tracheobronchial mucin
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
This MAb recognizes the peptide core of gastric mucin M1 (>1,000kDa) (recently identified as Mucin 5AC). Its epitope is destroyed by beta-mercaptoethanol and proteases but not by periodate treatment. Antibody to gastric mucin M1 reacts with the gastric epithelium of normal human gastrointestinal tract as well as with the precancerous and cancerous colon but not with normal adult colon. It also reacts with fetal colonic mucosa. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.