Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucochloric acid, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
87-56-9
C4H2Cl2O3
168.96
MFCD00006966
LUMLZKVIXLWTCI-IHWYPQMZSA-N
mucochloric acid,2,3-dichloromaleic aldehyde acid,kyselina mukochlorova,2z-2,3-dichloro-4-oxobut-2-enoic acid,2-butenoic acid, 2,3-dichloro-4-oxo-, 2z,aldehydodichloromaleic acid,dichloromalealdehydic acid,unii-5i5877jhiw,kyselina mukochlorova czech,ccris 6597
2771871
(Z)-2,3-dichloro-4-oxobut-2-enoic acid
C(=O)C(=C(C(=O)O)Cl)Cl
87-56-9
C4H2Cl2O3
168.96
MFCD00006966
LUMLZKVIXLWTCI-IHWYPQMZSA-N
mucochloric acid,2,3-dichloromaleic aldehyde acid,kyselina mukochlorova,2z-2,3-dichloro-4-oxobut-2-enoic acid,2-butenoic acid, 2,3-dichloro-4-oxo-, 2z,aldehydodichloromaleic acid,dichloromalealdehydic acid,unii-5i5877jhiw,kyselina mukochlorova czech,ccris 6597
2771871
(Z)-2,3-dichloro-4-oxobut-2-enoic acid
C(=O)C(=C(C(=O)O)Cl)Cl
Infrared Spectrum Authentic
Water 4% max. (K.F.)
Quantity 100g
Packaging Plastic bottle
Melting Point 125.0°C to 128.0°C
Color White to Beige
Flash Point 186°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC(CHO)=C(Cl)CO2H
Beilstein 03, 727
Merck Index 15, 6383
Solubility Information Solubility in water: soluble in hot water. Other solubilities: slightly soluble in cold water, soluble in hot benzene and alcohol
Formula Weight 168.96
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Mucochloric acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Toxic if swallowed.
May cause respiratory irritation.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Obtain special instructions before use.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger