Alfa Aesar™ Mucochloric acid, 99% (dry wt.), water <4.0%

Manufacturer: Alfa Aesar™ B2452322

Quantity 100g

 View more versions of this product

Catalog No. AAB2452322

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 87-56-9
    Molecular Formula C4H2Cl2O3
    Formula Weight 168.96
    MDL Number MFCD00006966
    UN Number UN2923
    Beilstein 1705641
    Merck Index 14,6297
    Synonym 2,3-Dichloromalealdehydic acid
    Chemical Name or Material Mucochloric acid

    Specifications

    Formula Weight 168.96
    Melting Point 125° to 128°C (dried)
    TSCA TSCA
    Molecular Formula C4H2Cl2O3
    CAS 87-56-9
    UN Number UN2923
    Beilstein 1705641
    Merck Index 14,6297
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 186°C (367°F)
    Quantity 100g
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    MDL Number MFCD00006966
    Synonym 2,3-Dichloromalealdehydic acid
    Health Hazard 2 GHS H Statement
    H301-H314-H318-H341
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing genetic defects.
    RTECS Number AS7700000
    Chemical Name or Material Mucochloric acid
    Assay 99%, (dry wt.), water <4.0%

    GHS H Statement
    H301-H314-H318-H341
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing genetic defects.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit